Anti-IgG2 beta Goat Polyclonal Antibody (CF™ 555)
Supplier: Biotium
Certificates
About this item
Type: Secondary
Antigen: IgG2 beta
Clonality: Polyclonal
Clone:
Conjugation: CF™ 555
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 71004-232
- 71004-230
- Antigen Symbol:
- IgG2 beta
- IgG2 beta
- Antigen Name:
- Immunoglobulin G2 beta
- Immunoglobulin G2 beta
- Antibody Type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- CF® 555
- CF® 555
- Reactivity:
- Mouse
- Mouse
- Host:
- Goat
- Goat
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Cross Adsorption:
- Minimal Cross-Reactivity: Bv, Hu, Rb
- Minimal Cross-Reactivity: Bv, Hu, Rb
Specifications
Frequently Bought Together