Order Entry
United States
ContactUsLinkComponent
Anti-IgG Donkey Polyclonal Antibody (CF™ 350)
Anti-IgG Donkey Polyclonal Antibody (CF™ 350)
Supplier:  Biotium
Certificates
undefined
Anti-IgG Donkey Polyclonal Antibody (CF™ 350)
Supplier:  Biotium

Specifications

  • Catalog No:
  • 89138-832
  • 89138-834
  • Antigen Symbol:
  • IgG
  • IgG
  • Antigen Name:
  • Immunoglobulin G
  • Immunoglobulin G
  • Antibody Type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • CF® 350
  • CF® 350
  • Reactivity:
  • Rabbit
  • Rabbit
  • Host:
  • Donkey
  • Donkey
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow Cytometry:
  • Yes
  • Yes
  • Epitope:
  • Heavy and Light chain
  • Heavy and Light chain
  • Cross Adsorption:
  • Minimal Cross-Reactivity: Bv, Ch, Gt, GP, Hs, Hu, Ms, Sh, SHm
  • Minimal Cross-Reactivity: Bv, Ch, Gt, GP, Hs, Hu, Ms, Sh, SHm

Specifications

Frequently Bought Together