Anti-CCK4 (PTK7) Rabbit Polyclonal Antibody
Catalog # 80084-428
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Symbol:CCK4 (PTK7)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:N-Terminal
- Cross Adsorption:No
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80084-428
- Supplier no.:AP7800A
Specifications
About this item
Type: Primary
Antigen: CCK4 (PTK7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human