About this item
Purified anti-β-Actin [Poly6221]; Isotype: Rabbit Polyclonal IgG; Reactivity: Mouse, Rat, Human; Apps: WB, IF, IHC; Size: 200 μl
Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 10 μl per 5 ml antibody dilution buffer for each mini-gel. For immunofluorescence microscopy: Use a dilution range of 1:100~1:400. For IHC, use a 1:100 dilution of antibody for staining. Antigen retrieval for IHC of formalin-fixed paraffin-embedded tissue using 0.01 M sodium citrate buffer is recommended. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: b-Actin
Clonality: Polyclonal
Clone: Poly6221
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit Polyclonal IgG
Reactivity: Mouse;Rat;Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 622102-BL
- 622101-BL
- Antigen Name:
- b-Actin
- b-Actin
- Antigen Synonyms:
- cytoplasmic 1|ACTB|Actin
- cytoplasmic 1|ACTB|Actin
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Poly6221
- Poly6221
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 11461
- 11461
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Peptide-KLH, NH2 terminus
- Peptide-KLH, NH2 terminus
- Form:
- This antibody is provided in phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 50% glycerol.
- This antibody is provided in phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 50% glycerol.
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.00 mg/mL
- 0.00 mg/mL
- Storage Temperature:
- Upon receipt, store frozen at -20 °C.
- Upon receipt, store frozen at -20 °C.
- Shipping Temperature:
- Ambient RT
- Ambient RT