To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:TCP-1beta
- Clonality:Monoclonal
- Clone:F39P7F11
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Frog,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Form:This antibody is provided in phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide at 0.5 mg/ml
- Gene ID:21454
- Antigen Synonyms:TCP1B|CCTB|T-complex protein 1 subunit beta|CCT beta
- Storage Temperature:Upon receipt, store undiluted between 2 °C and 8 °C.
- Concentration:0.50 mg/mL
- Shipping Temperature:Ambient RT
- Immunogen:Ovalbumin-conjugated synthetic Peptide RKRVPDHHPC
- Purification:The antibody was purified by affinity chromatography.
- Cat. No.:635502-BL
- Supplier no.:635502
Specifications
About this item
Purified anti-TCP-1β [F39P7F11]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat, Frog; Apps: WB; Size: 100 μg
Each lot of this antibody is quality control tested by Western blotting. For Western blotting, suggested working dilution(s): Use 5-10 μg per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: TCP-1beta
Clonality: Monoclonal
Clone: F39P7F11
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;Mouse;Rat;Frog