Order Entry
United States
ContactUsLinkComponent
Anti-TLR4 (CD284)/MD2 Complex Rat Monoclonal Antibody [clone: MTS510]
Anti-TLR4 (CD284)/MD2 Complex Rat Monoclonal Antibody [clone: MTS510]
Supplier:  Biolegend
Certificates
Anti-TLR4 (CD284)/MD2 Complex Rat Monoclonal Antibody [clone: MTS510]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • 117601-BL
  • 117602-BL
  • Antigen Symbol:
  • TLR4 (CD284)/MD2 Complex
  • TLR4 (CD284)/MD2 Complex
  • Antigen Synonyms:
  • TLR4|Toll-like Receptor 4/MD2 Complex|TLR4/MD2 Complex
  • TLR4|Toll-like Receptor 4/MD2 Complex|TLR4/MD2 Complex
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • MTS510
  • MTS510
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 21898
  • 21898
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Western Blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Ba/F3 cell line transfected with mouse TLR4 /MD2 complex
  • Ba/F3 cell line transfected with mouse TLR4 /MD2 complex
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Purification:
  • The antibody was purified by affinity chromatography.
  • The antibody was purified by affinity chromatography.
  • Concentration:
  • 0.50 mg/mL
  • 0.50 mg/mL
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications

Related Information

Frequently Bought Together