Order Entry
United States
ContactUsLinkComponent
Anti-TNF-a Rat Monoclonal Antibody [clone: 6B8]
Anti-TNF-a Rat Monoclonal Antibody [clone: 6B8]
Supplier:  Biolegend
Certificates
Anti-TNF-a Rat Monoclonal Antibody [clone: 6B8]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • 510802-BL
  • 510801-BL
  • Antigen Symbol:
  • TNF-a
  • TNF-a
  • Antigen Synonyms:
  • Differentiation inducing factor (DIF)|Necrosin|TNFSF2|TNFA|Cachectin|Tumor necrosis factor-α|Macrophage cytotoxic factor (MCF)
  • Differentiation inducing factor (DIF)|Necrosin|TNFSF2|TNFA|Cachectin|Tumor necrosis factor-α|Macrophage cytotoxic factor (MCF)
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 6B8
  • 6B8
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 21926
  • 21926
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Gel Shift:
  • Yes
  • Yes
  • Immunogen:
  • recombinant mouse TNF-α
  • recombinant mouse TNF-α
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Purification:
  • The antibody was purified by affinity chromatography.
  • The antibody was purified by affinity chromatography.
  • Concentration:
  • 0.50 mg/mL
  • 0.50 mg/mL
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications

Related Information

Frequently Bought Together