Order Entry
United States
ContactUsLinkComponent
Anti-GITR Ligand Rat Monoclonal Antibody [clone: YGL 386]
Anti-GITR Ligand Rat Monoclonal Antibody [clone: YGL 386]
  120302-BL
 :  Biolegend
Anti-GITR Ligand Rat Monoclonal Antibody [clone: YGL 386]
  120302-BL
 :  Biolegend
 :  120302
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Symbol:
    GITR Ligand
  • Clonality:
    Monoclonal
  • Clone:
    YGL 386
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Mouse
  • Size:
    200 μg
  • Environmentally Preferable:
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Gene ID:
    240873
  • Antigen Synonyms:
    TL6|AITRL|GITRL|TNFSF18
  • Storage Temperature:
    The antibody solution should be stored undiluted between 2 °C and 8 °C.
  • Concentration:
    0.50 mg/mL
  • Shipping Temperature:
    Ambient RT
  • Immunogen:
    Recombinant mouse GITRL protein and Transfected cells
  • Purification:
    The antibody was purified by affinity chromatography.
  • Cat. No.:
    120302-BL
  • Supplier no.:
    120302

 

 

Purified anti-mouse GITR Ligand [YGL 386]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: GITR Ligand
Clonality: Monoclonal
Clone: YGL 386
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse