Anti-Perforin Mouse Monoclonal Antibody [clone: dG9]
Catalog # 308102-BL
Supplier: Biolegend
Specifications
- Antibody Type:Primary
- Antigen Name:Perforin
- Clonality:Monoclonal
- Clone:dG9
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Human,Bovine
- Size:100 μg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Gene ID:5551
- Antigen Synonyms:lymphocyte pore-forming protein|cytolysin|lymphocyte pore forming protein|perforin-1|PRF1
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Concentration:0.50 mg/mL
- Shipping Temperature:Ambient RT
- Immunogen:Purified granules from the human lymphoma cell line
- Purification:The antibody was purified by affinity chromatography.
- Cat. No.:308102-BL
- Supplier no.:308102
Specifications
About this item
Purified anti-human Perforin [dG9]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Bovine (Cow, Cattle); Apps: ICFC, IP, IF, IHC; Size: 100 μg
Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.06 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: Perforin
Clonality: Monoclonal
Clone: dG9
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Cross-Reactivity: Bovine (Cow;Cattle)