Anti-SUSD2 Mouse Monoclonal Antibody [clone: W5C5]
Catalog # 327401-BL
Supplier: Biolegend
Specifications
- Antibody Type:Primary
- Antigen Symbol:SUSD2
- Clonality:Monoclonal
- Clone:W5C5
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Baboon,African green monkey,Cynomolgus monkey,Human
- Size:25 μg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Gene ID:56241
- Antigen Synonyms:Sushi domain containing 2 (SUSD2)|MSC (W5C5)|Mesenchymal Stem Cells (MSC)
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Concentration:0.50 mg/mL
- Shipping Temperature:Ambient RT
- Immunogen:Retinoblastoma cell line (Weri-RB-1 cells).
- Purification:The antibody was purified by affinity chromatography.
- Cat. No.:327401-BL
- Supplier no.:327401
Specifications
About this item
Purified anti-human SUSD2 [W5C5]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC, IF; Size: 25 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: SUSD2
Clonality: Monoclonal
Clone: W5C5
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus