Purified anti-human GM-CSF [BVD2-23B6]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: ELISA Capture, IP, WB; Size: 500 μg
Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 2-6 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of GM-CSF recombinant protein ranging from 500 to 4 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: GM-CSF
Clonality: Monoclonal
Clone: BVD2-23B6
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 502202-BL
- 502201-BL
- Antigen Symbol:
- GM-CSF
- GM-CSF
- Antigen Synonyms:
- Eo-CSF|CSF-α|BPA|Granulocyte/macrophage colony-stimulating factor|Pluripoietin-α|GMCSF1
- Eo-CSF|CSF-α|BPA|Granulocyte/macrophage colony-stimulating factor|Pluripoietin-α|GMCSF1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- BVD2-23B6
- BVD2-23B6
- Reactivity:
- Human
- Human
- Host:
- Rat
- Rat
- Gene ID:
- 1437
- 1437
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Gel Shift:
- Yes
- Yes
- Immunogen:
- E. coli -expressed, recombinant human GM-CSF
- E. coli -expressed, recombinant human GM-CSF
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Shipping Temperature:
- Ambient RT
- Ambient RT