Anti-CD75 Mouse Monoclonal Antibody [clone: LN1]
Catalog # 326901-BL
Supplier: Biolegend
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD75
- Clonality:Monoclonal
- Clone:LN1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgM kappa
- Reactivity:Human
- Western Blot:Yes
- Size:25 μg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Gene ID:6480
- Antigen Synonyms:B cell antigen CD75|Beta-galactoside alpha-2,6-sialyltransferase 1 (ST6GAL1)
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Concentration:0.50 mg/mL
- Shipping Temperature:Ambient RT
- Immunogen:Human PBL nuclei
- Purification:This antibody is at >85% purity.
- Cat. No.:326901-BL
- Supplier no.:326901
Specifications
About this item
Purified anti-human CD75 [LN1]; Isotype: Mouse IgM, κ; Reactivity: Human; Apps: FC, IHC, WB; Size: 25 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per 106 cells in 100 μl volume or 100 μl whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD75
Clonality: Monoclonal
Clone: LN1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgM, κ
Reactivity: Human