Anti-CD286 (TLR6) Mouse Monoclonal Antibody [clone: TLR6.127]
Catalog # 334702-BL
Supplier: Biolegend
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD286 (TLR6)
- Clonality:Monoclonal
- Clone:TLR6.127
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 μg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Gene ID:10333
- Antigen Synonyms:TLR6
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Concentration:0.50 mg/mL
- Shipping Temperature:Ambient RT
- Immunogen:Human TLR6 transfected Ba/F3 cells
- Purification:The antibody was purified by affinity chromatography.
- Cat. No.:334702-BL
- Supplier no.:334702
Specifications
About this item
Purified anti-human CD286 (TLR6) [TLR6.127]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD286 (TLR6)
Clonality: Monoclonal
Clone: TLR6.127
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human