Anti-CD275 (B7-H2, B7-RP1, ICOSL) Mouse Monoclonal Antibody [clone: 9F.8A4]
Catalog # 329801-BL
Supplier: Biolegend
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD275 (B7-H2, B7-RP1, ICOSL)
- Clonality:Monoclonal
- Clone:9F.8A4
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:25 μg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Gene ID:23308
- Antigen Synonyms:B7-H2|B7-RP1|ICOSL|B7H2
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Concentration:0.50 mg/mL
- Shipping Temperature:Ambient RT
- Immunogen:Human B7H2-mIg fusion protein
- Purification:The antibody was purified by affinity chromatography.
- Cat. No.:329801-BL
- Supplier no.:329801
Specifications
About this item
Purified anti-human CD275 (B7-H2, B7-RP1, ICOSL) [9F.8A4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD275 (B7-H2, B7-RP1, ICOSL)
Clonality: Monoclonal
Clone: 9F.8A4
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human