About this item
Purified anti-human CD274 (B7-H1, PD-L1) [29E.2A3]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, IHC-P; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. For immunohistochemical staining on formalin-fixed paraffin-embedded tissue sections, the suggested use of this reagent is 5.0 - 10 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD274 (B7-H1, PD-L1)
Clonality: Monoclonal
Clone: 29E.2A3
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Specifications
- Catalog No:
- 329701-BL
- 329702-BL
- Antigen Symbol:
- CD274 (B7-H1, PD-L1)
- CD274 (B7-H1, PD-L1)
- Antigen Synonyms:
- PDL1|Programmed cell death ligand 1 (PD-L1)|29E2A3|B7 homolog 1 (B7-H1) B7H1|CD274|Immune checkpoints
- PDL1|Programmed cell death ligand 1 (PD-L1)|29E2A3|B7 homolog 1 (B7-H1) B7H1|CD274|Immune checkpoints
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 29E.2A3
- 29E.2A3
- Reactivity:
- Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Human
- Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 29126
- 29126
- Isotype:
- IgG2b kappa
- IgG2b kappa
- ImmunoChemistry:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Full length human PD-L1
- Full length human PD-L1
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Shipping Temperature:
- Ambient RT
- Ambient RT