Purified anti-c-Myc [9E10]; Isotype: Mouse IgG1; Reactivity: Human and fusion protein (EQKLISEEDL tag epitope) in all species; Apps: ELISA, IF, IHC, IP, WB; Size: 100 μg
Each lot of this antibody is quality control tested by Western blotting. For Western blotting, suggested working dilution(s): Use 5 μg per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: c-Myc
Clonality: Monoclonal
Clone: 9E10
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human and fusion protein (EQKLISEEDL tag epitope) in all species
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 626802-BL
- 626801-BL
- Antigen Symbol:
- c-Myc
- c-Myc
- Antigen Synonyms:
- Myc proto-oncogene protein|Oncogene Myc
- Myc proto-oncogene protein|Oncogene Myc
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 9E10
- 9E10
- Reactivity:
- Multiple
- Multiple
- Host:
- Mouse
- Mouse
- Gene ID:
- 4609
- 4609
- Isotype:
- IgG1
- IgG1
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Gel Shift:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Amino acids 408-439, C-terminal region of human c-myc
- Amino acids 408-439, C-terminal region of human c-myc
- Form:
- This antibody is provided in phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide at 0.5 mg/ml.
- This antibody is provided in phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide at 0.5 mg/ml.
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage Temperature:
- Upon receipt, store undiluted between 2 °C and 8 °C.
- Upon receipt, store undiluted between 2 °C and 8 °C.
- Shipping Temperature:
- Ambient RT
- Ambient RT