Anti-c-Fos Rabbit Polyclonal Antibody [clone: Poly6414]
Supplier: Biolegend
Certificates
About this item
Purified anti-c-Fos [Poly6414]; Isotype: Rabbit Polyclonal IgG; Reactivity: Human, Mouse, Rat; Apps: WB; Size: 100 μl
Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 10 μl per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: c-Fos
Clonality: Polyclonal
Clone: Poly6414
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit Polyclonal IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 641402-BL
- 641401-BL
- Antigen Symbol:
- c-Fos
- c-Fos
- Antigen Synonyms:
- Oncogene Fos|FBJ murine viral (v-fos) oncogene homolog
- Oncogene Fos|FBJ murine viral (v-fos) oncogene homolog
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Poly6414
- Poly6414
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 2353
- 2353
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Form:
- This antibody is provided in phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 50% glycerol.
- This antibody is provided in phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 50% glycerol.
- Purification:
- The antibody was purified by antigen-affinity chromatography.
- The antibody was purified by antigen-affinity chromatography.
- Concentration:
- 0.00 mg/mL
- 0.00 mg/mL
- Storage Temperature:
- Upon receipt, store at -20 °C.
- Upon receipt, store at -20 °C.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together