Anti-CD1d (CD1.1, Ly-38) Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: 1B1]
Supplier: Biolegend
Certificates
About this item
PerCP/Cy5.5 anti-mouse CD1d (CD1.1, Ly-38) [1B1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
* PerCP/Cy5.5 has a maximum absorption of 482 nm and a maximum emission of 690 nm.
Type: Primary
Antigen: CD1d (CD1.1, Ly-38)
Clonality: Monoclonal
Clone: 1B1
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Specifications
- Catalog No:
- 123514-BL
- 123513-BL
- Antigen Symbol:
- CD1d (CD1.1, Ly-38)
- CD1d (CD1.1, Ly-38)
- Antigen Synonyms:
- CD1D|Ly-38|CD1.1
- CD1D|Ly-38|CD1.1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- Clone:
- 1B1
- 1B1
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 12479
- 12479
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Mouse Cd1.1 cDNA-transfected RMA-S mouse T lymphoma
- Mouse Cd1.1 cDNA-transfected RMA-S mouse T lymphoma
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with PerCP/Cy5.5 under optimal conditions. The solution is free of unconjugated PerCP/Cy5.5 and unconjugated antibody.
- The antibody was purified by affinity chromatography, and conjugated with PerCP/Cy5.5 under optimal conditions. The solution is free of unconjugated PerCP/Cy5.5 and unconjugated antibody.
- Concentration:
- 0.20 mg/mL
- 0.20 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together