Order Entry
United States
ContactUsLinkComponent
Anti-CD38 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: HIT2]
Anti-CD38 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: HIT2]
Supplier:  Biolegend
Certificates
Anti-CD38 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: HIT2]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • 303519-BL
  • 303520-BL
  • Antigen Symbol:
  • CD38
  • CD38
  • Antigen Synonyms:
  • T10|ADP-ribosyl cyclase
  • T10|ADP-ribosyl cyclase
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PerCP (Peridinin-Chlorophyll Protein Complex)
  • PerCP (Peridinin-Chlorophyll Protein Complex)
  • Clone:
  • HIT2
  • HIT2
  • Reactivity:
  • Horse, Chimpanzee, Human, Bovine
  • Horse, Chimpanzee, Human, Bovine
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 952
  • 952
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography, and conjugated with PerCP under optimal conditions. The solution is free of unconjugated PerCP and unconjugated antibody.
  • The antibody was purified by affinity chromatography, and conjugated with PerCP under optimal conditions. The solution is free of unconjugated PerCP and unconjugated antibody.
  • Storage Temperature:
  • The CD38 antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The CD38 antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications

Related Information

Frequently Bought Together