Order Entry
United States
ContactUsLinkComponent
Anti-IgG2a, k Isotype Ctrl Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: RTK2758]
Anti-IgG2a, k Isotype Ctrl Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: RTK2758]
Supplier:  Biolegend
Certificates
Anti-IgG2a, k Isotype Ctrl Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: RTK2758]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • 400522-BL
  • 400521-BL
  • Antigen Symbol:
  • IgG2a k isotype ctrl
  • IgG2a k isotype ctrl
  • Antibody Type:
  • Control
  • Control
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)/Cy7®
  • PE (Phycoerythrin)/Cy7®
  • Clone:
  • RTK2758
  • RTK2758
  • Host:
  • Rat
  • Rat
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Trinitrophenol + KLH
  • Trinitrophenol + KLH
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Purification:
  • The immunoglobulin was purified by affinity chromatography, and conjugated with PE/Cy7 under optimal conditions. The solution is free of unconjugated PE/Cy7 and unconjugated immunoglobulin.
  • The immunoglobulin was purified by affinity chromatography, and conjugated with PE/Cy7 under optimal conditions. The solution is free of unconjugated PE/Cy7 and unconjugated immunoglobulin.
  • Concentration:
  • 0.20 mg/mL
  • 0.20 mg/mL
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications

Related Information

Frequently Bought Together