Anti-IgG1, k Isotype Ctrl Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: MOPC-21]
Supplier: Biolegend
Certificates
About this item
PE/Cy5 Mouse IgG1, κ Isotype Ctrl [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 100 tests
Each lot of this mouse IgG1, κ isotype control antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, use the isotype control at the same concentration as your primary antibody. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.
Type: Control
Antigen: IgG1, k Isotype Ctrl
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse
Specifications
- Catalog No:
- 400116-BL
- 400115-BL
- 400118-BL
- 400117-BL
- Antigen Symbol:
- IgG1 k isotype ctrl
- IgG1 k isotype ctrl
- IgG1 k isotype ctrl
- IgG1 k isotype ctrl
- Antibody Type:
- Control
- Control
- Control
- Control
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)/Cy5®
- PE (Phycoerythrin)/Cy5®
- PE (Phycoerythrin)/Cy5®
- PE (Phycoerythrin)/Cy5®
- Clone:
- MOPC-21
- MOPC-21
- MOPC-21
- MOPC-21
- Reactivity:
- Mouse
- Mouse
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa
- Flow Cytometry:
- Yes
- Yes
- Yes
- Yes
- Form:
- test sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA). µg sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- test sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA). µg sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- test sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA). µg sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- test sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA). µg sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with PE/Cy5 under optimal conditions. The solution is free of unconjugated PE/Cy5 and unconjugated antibody.
- The antibody was purified by affinity chromatography, and conjugated with PE/Cy5 under optimal conditions. The solution is free of unconjugated PE/Cy5 and unconjugated antibody.
- The antibody was purified by affinity chromatography, and conjugated with PE/Cy5 under optimal conditions. The solution is free of unconjugated PE/Cy5 and unconjugated antibody.
- The antibody was purified by affinity chromatography, and conjugated with PE/Cy5 under optimal conditions. The solution is free of unconjugated PE/Cy5 and unconjugated antibody.
- Concentration:
- 0.20 mg/mL
- 0.20 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together