Anti-CD117 (c-kit) Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 2B8]
PE/Cy5 anti-mouse CD117 (c-kit) [2B8]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD117 (c-kit)
Clonality: Monoclonal
Clone: 2B8
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
- Catalog No:
- 105810-BL
- 105809-BL
- Antigen Symbol:
- CD117 (c-kit)
- CD117 (c-kit)
- Antigen Synonyms:
- KIT|c-KIT|Stem Cell Factor Receptor (SCFR)
- KIT|c-KIT|Stem Cell Factor Receptor (SCFR)
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)/Cy5®
- PE (Phycoerythrin)/Cy5®
- Clone:
- 2B8
- 2B8
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 16590
- 16590
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Mouse bone marrow mast cells
- Mouse bone marrow mast cells
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with PE/Cy5 under optimal conditions. The solution is free of unconjugated PE/Cy5 and unconjugated antibody.
- The antibody was purified by affinity chromatography, and conjugated with PE/Cy5 under optimal conditions. The solution is free of unconjugated PE/Cy5 and unconjugated antibody.
- Concentration:
- 0.20 mg/mL
- 0.20 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Frequently Bought Together