Order Entry
United States
ContactUsLinkComponent
 
Anti-CD4 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: RPA-T4]
 :  Biolegend
 
Anti-CD4 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: RPA-T4]
 :  Biolegend

 

  • Catalog No:
  • 300509-BL
  • 300510-BL
  • Antigen Symbol:
  • CD4
  • CD4
  • Antigen Synonyms:
  • T4
  • T4
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)/Cy5®
  • PE (Phycoerythrin)/Cy5®
  • Clone:
  • RPA-T4
  • RPA-T4
  • Reactivity:
  • Cynomolgus monkey, Chimpanzee, Human
  • Cynomolgus monkey, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 920
  • 920
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography, and conjugated with PE/Cy5 under optimal conditions. The solution is free of unconjugated PE/Cy5 and unconjugated antibody.
  • The antibody was purified by affinity chromatography, and conjugated with PE/Cy5 under optimal conditions. The solution is free of unconjugated PE/Cy5 and unconjugated antibody.
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

 

Frequently Bought Together