Anti-IgG2c, k Isotype Ctrl Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RTK4174]
Catalog # 400707-BL
Supplier: Biolegend
Specifications
- Antibody Type:Control
- Antigen Symbol:IgG2c k isotype ctrl
- Clonality:Monoclonal
- Clone:RTK4174
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2c kappa
- Reactivity:Rat
- Size:25 μg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Concentration:0.20 mg/mL
- Shipping Temperature:Ambient RT
- Immunogen:Trinitrophenol + KLH
- Purification:The immunoglobulin was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated immunoglobulin.
- Cat. No.:400707-BL
- Supplier no.:400707
Specifications
About this item
PE Rat IgG2c, κ Isotype Ctrl [RTK4174]; Isotype: Rat IgG2c, κ; Reactivity: Rat; Apps: FC, ICFC; Size: 25 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.
Type: Control
Antigen: IgG2c, k Isotype Ctrl
Clonality: Monoclonal
Clone: RTK4174
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2c, κ
Reactivity: Rat