Anti-IgG (minimal x-reactivity) Goat Polyclonal Antibody (PE (Phycoerythrin)) [clone: Poly4053]
Catalog # 405307-BL
Supplier: Biolegend
Specifications
- Antibody Type:Primary
- Antigen Symbol:IgG (minimal x-reactivity)
- Clonality:Polyclonal
- Clone:Poly4053
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Mouse
- Size:200 μg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide
- Gene ID:16059
- Antigen Synonyms:IgG
- Storage Temperature:The IgG antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Concentration:0.20 mg/mL
- Shipping Temperature:Ambient RT
- Purification:The IgG antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- Cat. No.:405307-BL
- Supplier no.:405307
Specifications
About this item
PE Goat anti-mouse IgG (minimal x-reactivity) [Poly4053]; Isotype: Goat Polyclonal IgG; Reactivity: Mouse; Apps: FC; Size: 200 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: IgG (minimal x-reactivity)
Clonality: Polyclonal
Clone: Poly4053
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: Goat Polyclonal IgG
Reactivity: Mouse