Anti-AVPR1A Rabbit Polyclonal Antibody
Catalog # 89519-858
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:AVP Receptor V1a
- Antigen Symbol:AVPR1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal
- Gene ID:552
- Antigen Synonyms:AVPR1|AVPR V1a|Vascular/hepatic-type arginine vasopressin receptor|Vasopressin V1a receptor|Antidiuretic hormone receptor 1a|AVPR1A
- Amino Acid Number:355 to 383
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:46.8 kDa
- Sequence:KEKFNKEDTDSMSRC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping Temperature:4 °C
- Immunogen:This AVPR1A antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 355-383 amino acids from the C-terminal region of human AVPR1A.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89519-858
- Supplier no.:AP16568B
Specifications
About this item
Western Blot: 1:1000
Type: Primary
Antigen: AVPR1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human