Anti-CD300f (MAIR-V) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: TX70]
Catalog # 132704-BL
Supplier: Biolegend
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD300f (MAIR-V)
- Clonality:Monoclonal
- Clone:TX70
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Size:100 μg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Gene ID:246746
- Antigen Synonyms:CD300LF|CMRF35-like molecule 1|Myeloid-associated immunoglobulin-like receptor 5|CD300F|CD300f
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Concentration:0.20 mg/mL
- Shipping Temperature:Ambient RT
- Immunogen:MAIR-V transfectant (BW5147)
- Purification:The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- Cat. No.:132704-BL
- Supplier no.:132704
Specifications
About this item
PE anti-mouse MAIR-V [TX70]; Isotype: Rat IgG2a; Reactivity: Mouse; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.015 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD300f (MAIR-V)
Clonality: Monoclonal
Clone: TX70
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2a
Reactivity: Mouse