Order Entry
United States
ContactUsLinkComponent
Anti-B7-DC (CD273, PD-L2, PDL2, B7DC) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: TY25]
Anti-B7-DC (CD273, PD-L2, PDL2, B7DC) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: TY25]
Supplier:  Biolegend
Certificates
Anti-B7-DC (CD273, PD-L2, PDL2, B7DC) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: TY25]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • 107205-BL
  • 107206-BL
  • Antigen Symbol:
  • B7-DC (CD273, PD-L2, PDL2, B7DC)
  • B7-DC (CD273, PD-L2, PDL2, B7DC)
  • Antigen Synonyms:
  • B7DC|PDL2
  • B7DC|PDL2
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • TY25
  • TY25
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 58205
  • 58205
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Mouse B7-DC transfected cell line
  • Mouse B7-DC transfected cell line
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Purification:
  • The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
  • The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
  • Concentration:
  • 0.20 mg/mL
  • 0.20 mg/mL
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications

Related Information

Frequently Bought Together