Order Entry
United States
ContactUsLinkComponent
Anti-CD255 (TWEAK) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: CARL-1]
Anti-CD255 (TWEAK) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: CARL-1]
  308305-BL
 :  Biolegend
Anti-CD255 (TWEAK) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: CARL-1]
  308305-BL
 :  Biolegend
 :  308305
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Symbol:
    CD255 (TWEAK)
  • Clonality:
    Monoclonal
  • Clone:
    CARL-1
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG3 kappa
  • Reactivity:
    Baboon,
    African green monkey,
    Human
  • Size:
    25 tests
  • Environmentally Preferable:
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Gene ID:
    8742
  • Antigen Synonyms:
    TNF-like weak inducer of apoptosis|Apo-3 ligand|TNFSF12|DR3-L|TNFSF-12
  • Storage Temperature:
    The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
    Ambient RT
  • Immunogen:
    hTWEAK/2PK-3 transfectant cells
  • Purification:
    The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
  • Cat. No.:
    308305-BL
  • Supplier no.:
    308305

 

 

PE anti-human CD255 (TWEAK) [CARL-1]; Isotype: Mouse IgG3, κ; Reactivity: Human, African Green, Baboon; Apps: FC; Size: 25 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: CD255 (TWEAK)
Clonality: Monoclonal
Clone: CARL-1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG3, κ
Reactivity: Human;African Green;Baboon