PE anti-human CD158b (KIR2DL2/L3, NKAT2) [DX27]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.
Type: Primary
Antigen: CD158b (KIR2DL2/L3, NKAT2)
Clonality: Monoclonal
Clone: DX27
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
- Catalog No:
- 312605-BL
- 312606-BL
- Antigen Symbol:
- CD158b (KIR2DL2/L3, NKAT2)
- CD158b (KIR2DL2/L3, NKAT2)
- Antigen Synonyms:
- KIR2DL2/L3|CD158j (KIR2DS2|CD158b1 (KIR2DL2|CD158b2 (KIR2DL3|KIR-NKAT2|p58.3)|p50.2)|p58.2)
- KIR2DL2/L3|CD158j (KIR2DS2|CD158b1 (KIR2DL2|CD158b2 (KIR2DL3|KIR-NKAT2|p58.3)|p50.2)|p58.2)
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- DX27
- DX27
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 3803
- 3803
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Flow Cytometry:
- Yes
- Yes
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT