Anti-IgG2b, k Isotype Ctrl Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: MPC-11]
Catalog # 400331-BL
Supplier: Biolegend
Specifications
- Antibody Type:Control
- Antigen Symbol:IgG2b k isotype ctrl
- Clonality:Monoclonal
- Clone:MPC-11
- Conjugation:PB (Pacific Blue)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Size:100 μg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Concentration:0.50 mg/mL
- Shipping Temperature:Ambient RT
- Purification:The immunoglobulin was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™.
- Cat. No.:400331-BL
- Supplier no.:400331
Specifications
About this item
Pacific Blue™ Mouse IgG2b, κ Isotype Ctrl [MPC-11]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest. * Pacific Blue™ has a maximum emission of 455 nm when it is excited at 405 nm. Prior to using Pacific Blue™ conjugate for flow cytometric analysis, please verify your flow cytometer's capability of exciting and detecting the fluorochrome.
Type: Control
Antigen: IgG2b, k Isotype Ctrl
Clonality: Monoclonal
Clone: MPC-11
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Mouse