Specifications
- Antibody Type:Primary
- Antigen Symbol:IgG (minimal x-reactivity)
- Clonality:Polyclonal
- Clone:Poly4053
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Gel Shift:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:500 μl
- Form:Aqueous buffered solution containing a carrier protein and preservatives.
- Gene ID:16059
- Antigen Synonyms:IgG
- Concentration:0.00
- Shipping Temperature:Ambient RT
- Purification:The polyclonal antibody was purified from goat antiserum by mouse immunoglobulin affinity chromatography.
- Cat. No.:405306-BL
- Supplier no.:405306
Specifications
About this item
HRP Goat anti-mouse IgG (minimal x-reactivity) [Poly4053]; Isotype: Goat Polyclonal IgG; Reactivity: Mouse; Apps: ELISA, IHC, WB; Size: 500 μl
Each lot of this antibody is quality control tested by ELISA assay. Do not use this product beyond the expiration date. For ELISA or Western blot analysis, the reagent should be titrated between 1:2,000 - 1:5,000 to determine optimal conditions. For IHC on tissue sections, the reagent should be titrated between 1:500 - 1:2000 to determine optimal conditions. It is recommended that the reagent be titrated for optimal performance for each application. Avoid using diluent solutions containg sodium azide. Sodium azide is an inhibitor of horseradish peroxidase.
Type: Primary
Antigen: IgG (minimal x-reactivity)
Clonality: Polyclonal
Clone: Poly4053
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: Goat Polyclonal IgG
Reactivity: Mouse