About this item
FITC anti-human FcεRIα [AER-37 (CRA-1)]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.
Type: Primary
Antigen: Fc epsilon RI alpha
Clonality: Monoclonal
Clone: AER-37 (CRA-1)
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Baboon;Cynomolgus;Rhesus
Specifications
- Catalog No:
- 334607-BL
- 334608-BL
- Antigen Symbol:
- Fc epsilon RI alpha
- Fc epsilon RI alpha
- Antigen Synonyms:
- FceRI-a|FceRIa|FceRI alpha|FceRI-alpha|high affinity IgE receptorBasophilEosinophilMast CellPlateletsFcεRIα
- FceRI-a|FceRIa|FceRI alpha|FceRI-alpha|high affinity IgE receptorBasophilEosinophilMast CellPlateletsFcεRIα
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- AER-37 (CRA-1)
- AER-37 (CRA-1)
- Reactivity:
- Baboon, Cynomolgus monkey, Rhesus monkey, Human
- Baboon, Cynomolgus monkey, Rhesus monkey, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 2205
- 2205
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow Cytometry:
- Yes
- Yes
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
- The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT