Anti-MOCS3 Rabbit Polyclonal Antibody
Catalog # 89514-996
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Molybdenum Cofactor Synthesis 3
- Antigen Symbol:MOCS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Mouse
- Western Blot:Yes
- Size:400 µL
- Epitope:N-Terminal
- Gene ID:27304
- Antigen Synonyms:UBA4
- Amino Acid Number:40 to 66
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:49.669 kDa
- Sequence:LSRDEILRYSRQC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping Temperature:4 °C
- Immunogen:This MOCS3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 40-66 amino acids from the N-terminal region of human MOCS3.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89514-996
- Supplier no.:AP19278A
Specifications
About this item
Western Blot: 1:1000
Type: Primary
Antigen: MOCS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse