Biotin anti-mouse Ly-6G/Ly-6C (Gr-1) [RB6-8C5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IHC, IP, WB; Size: 500 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: Ly-6G/Ly-6C (Gr-1)
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 108403-BL
- 108404-BL
- Antigen Symbol:
- Ly-6G/Ly-6C (Gr-1)
- Ly-6G/Ly-6C (Gr-1)
- Antigen Synonyms:
- Ly6G|LY6G|Gr-1|Ly6C
- Ly6G|LY6G|Gr-1|Ly6C
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- RB6-8C5
- RB6-8C5
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 17067, 546644
- 17067, 546644
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Raised against granulocytes of mouse origin
- Raised against granulocytes of mouse origin
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with biotin under optimal conditions. The solution is free of unconjugated biotin.
- The antibody was purified by affinity chromatography, and conjugated with biotin under optimal conditions. The solution is free of unconjugated biotin.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage Temperature:
- The Ly-6G/Ly-6C antibody solution should be stored undiluted between 2 °C and 8 °C. Do not freeze.
- The Ly-6G/Ly-6C antibody solution should be stored undiluted between 2 °C and 8 °C. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT