Anti-Allophycocyanin (APC) Mouse Monoclonal Antibody (Biotin) [clone: APC003]
Biotin anti-Allophycocyanin (APC) [APC003]; Isotype: Mouse IgG1, κ; Reactivity: 2nd Step; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Use at concentrations comparable to those of the primary antibody of interest. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: Allophycocyanin (APC)
Clonality: Monoclonal
Clone: APC003
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: 2nd Step
- Catalog No:
- 408004-BL
- 408003-BL
- Antigen Symbol:
- APC
- APC
- Antigen Name:
- Allophycocyanin
- Allophycocyanin
- Antigen Synonyms:
- DP2.5|APC|FAP|FPC|GS|DP3|DP2|Adenomatous Polyposis Coli
- DP2.5|APC|FAP|FPC|GS|DP3|DP2|Adenomatous Polyposis Coli
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- APC003
- APC003
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- allophycocyanin
- allophycocyanin
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with biotin under optimal conditions. The solution is free of unconjugated biotin.
- The antibody was purified by affinity chromatography, and conjugated with biotin under optimal conditions. The solution is free of unconjugated biotin.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Frequently Bought Together