Anti-IgM, k Isotype Ctrl Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MM-30]
APC Mouse IgM, κ Isotype Ctrl [MM-30]; Isotype: Mouse IgM, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 100 tests
Each lot of this mouse IgM, κ isotype control antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, use the isotype control at the same concentration as your primary antibody. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.
Type: Control
Antigen: IgM, k Isotype Ctrl
Clonality: Monoclonal
Clone: MM-30
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgM, κ
Reactivity: Mouse
- Catalog No:
- 401613-BL
- 401616-BL
- Antigen Symbol:
- IgM k isotype ctrl
- IgM k isotype ctrl
- Antibody Type:
- Control
- Control
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- MM-30
- MM-30
- Reactivity:
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgM kappa
- IgM kappa
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Trinitrophenol + KLH
- Trinitrophenol + KLH
- Form:
- test size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
µg size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide. - test size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA). µg size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was conjugated with APC under optimal conditions, and is at >85% purity. The solution is free of unconjugated APC.
- The antibody was conjugated with APC under optimal conditions, and is at >85% purity. The solution is free of unconjugated APC.
- Concentration:
- 0.20 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Frequently Bought Together