Anti-I-A/I-E Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: M5/114.15.2]
Supplier: Biolegend
Certificates
About this item
APC anti-mouse I-A/I-E [M5/114.15.2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining,the suggested use of this reagent is 0.25 μg per 106cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: I-A/I-E
Clonality: Monoclonal
Clone: M5/114.15.2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Specifications
- Catalog No:
- 107613-BL
- 107614-BL
- Antigen Symbol:
- I-A/I-E
- I-A/I-E
- Antigen Synonyms:
- MHC Class II
- MHC Class II
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- M5/114.15.2
- M5/114.15.2
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 14961, 14969
- 14961, 14969
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Activated C57BL/6 mouse spleen cells
- Activated C57BL/6 mouse spleen cells
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
- The antibody was purified by affinity chromatography, and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
- Concentration:
- 0.20 mg/mL
- 0.20 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together