Anti-B3GALT2 Rabbit Polyclonal Antibody
Catalog # 89521-050
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 2
- Antigen Symbol:B3GALT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Mouse
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal
- Gene ID:8707
- Antigen Synonyms:beta3Gal-T2|BETA3GALT2|GLCT2
- Amino Acid Number:394 to 422
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:49.213 kDa
- Sequence:CAKEKAGRYRHRKLH
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.35mg/ml
- Shipping Temperature:4 °C
- Immunogen:This B3GALT2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 394-422 amino acids from the C-terminal region of human B3GALT2.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89521-050
- Supplier no.:AP14494B
Specifications
About this item
Western Blot: 1:1000
Type: Primary
Antigen: B3GALT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse