Order Entry
United States
ContactUsLinkComponent
Anti-CD56 (NCAM) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-188]
Anti-CD56 (NCAM) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-188]
Catalog # 304610-BL
Supplier:  Biolegend
Anti-CD56 (NCAM) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-188]
Catalog # 304610-BL
Supplier:  Biolegend
Supplier Number:  304610
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    CD56 (NCAM)
  • Clonality:
    Monoclonal
  • Clone:
    MEM-188
  • Conjugation:
    APC (Allophycocyanin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human,
    Bovine
  • Size:
    100 tests
  • Environmentally Preferable:
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Gene ID:
    4684
  • Antigen Synonyms:
    NCAM|NCAM1
  • Storage Temperature:
    The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
    Ambient RT
  • Immunogen:
    KG-1 human acute myelogenous leukemia cell line
  • Purification:
    The antibody was purified by affinity chromatography, and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
  • Cat. No.:
    304610-BL
  • Supplier no.:
    304610

Specifications

About this item

APC anti-human CD56 (NCAM) [MEM-188]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Cattle (Bovine, Cow); Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: CD56 (NCAM)
Clonality: Monoclonal
Clone: MEM-188
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;Cross-Reactivity: Cattle (Bovine;Cow)