About this item
APC anti-human CD4 [RPA-T4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee, Cynomolgus; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining using the μg size, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Chimpanzee;Cynomolgus
Specifications
- Catalog No:
- 300514-BL
- 10757-226
- 10757-206
- Antigen Symbol:
- CD4
- CD4
- CD4
- Antigen Synonyms:
- T4
- T4
- T4
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- RPA-T4
- RPA-T4
- RPA-T4
- Reactivity:
- Cynomolgus monkey, Chimpanzee, Human
- Cynomolgus monkey, Chimpanzee, Human
- Cynomolgus monkey, Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 920
- 920
- 920
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa
- Flow Cytometry:
- Yes
- Form:
- µg size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide. Test sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- µg size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide. Test sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- µg size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide. Test sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
- The antibody was purified by affinity chromatography, and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
- The antibody was purified by affinity chromatography, and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
- Concentration:
- 0.20 mg/mL
- Storage Temperature:
- The CD4 antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The CD4 antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The CD4 antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
- Ambient RT