Anti-TMBIM1 Rabbit Polyclonal Antibody
RL200-401-A57
- Antibody Type:Primary
- Antigen Name:Transmembrane BAX Inhibitor Motif Containing 1
- Antigen Symbol:TMBIM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:64114
- Antigen Synonyms:MSTP100|RECS1|LFG3|PP1201|MST100
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage Temperature:–20 °C
- Concentration:1.2 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:This Protein A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a recombinant Protein corresponding to the amino terminus of human TMBIM1 Protein.
- Purification:Protein A purified
- Cat. No.:RL200-401-A57
- Supplier no.:200-401-A57
Type: Primary
Antigen: TMBIM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human