To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:CD3/(CD16+CD56)
- Clonality:Monoclonal
- Clone:UCHT1/3G8/MEM-188
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a,IgG1
- Reactivity:Human
- Size:50 tests
- Environmentally Preferable:
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Gene ID:920
- Antigen Synonyms:CD3|CD3/(CD16+CD56)
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Concentration:0.00
- Shipping Temperature:Ambient RT
- Purification:This reagent is a combination of FITC conjugated UCHT1 and PE conjugated 3G8 and MEM-188 in optimal concentration for two-color flow cytometric analysis.
- Cat. No.:319101-BL
- Supplier no.:319101
Anti-human CD3 FITC/(CD16+CD56) PE Cocktail [UCHT1/3G8/MEM-188]; Isotype: Mouse IgG1/Mouse IgG2a; Reactivity: Human; Apps: FC; Size: 50 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 20 μl per million cells or 20 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD3/(CD16+CD56)
Clonality: Monoclonal
Clone: UCHT1/3G8/MEM-188
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1/Mouse IgG2a
Reactivity: Human