Order Entry
United States
ContactUsLinkComponent
Anti-IgG2b, k Isotype Ctrl Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: MPC-11]
Anti-IgG2b, k Isotype Ctrl Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: MPC-11]
Catalog # 400334-BL
Supplier:  Biolegend
Anti-IgG2b, k Isotype Ctrl Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: MPC-11]
Catalog # 400334-BL
Supplier:  Biolegend
Supplier Number:  400334

Specifications

  • Antibody Type:
    Control
  • Antigen Symbol:
    IgG2b k isotype ctrl
  • Clonality:
    Monoclonal
  • Clone:
    MPC-11
  • Conjugation:
    Alexa Fluor® 700
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Mouse
  • Size:
    100 μg
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Storage Temperature:
    The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Concentration:
    0.50 mg/mL
  • Shipping Temperature:
    Ambient RT
  • Purification:
    The immunoglobulin was purified by affinity chromatography, and conjugated with Alexa Fluor® 700 under optimal conditions.
  • Cat. No.:
    400334-BL
  • Supplier no.:
    400334

Specifications

About this item

Alexa Fluor® 700 Mouse IgG2b, κ Isotype Ctrl [MPC-11]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 100 μg

This reagent is developed for immunofluorescent staining for flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Control
Antigen: IgG2b, k Isotype Ctrl
Clonality: Monoclonal
Clone: MPC-11
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Mouse