Order Entry
United States
ContactUsLinkComponent
Anti-TRA-1-60-R Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: TRA-1-60-R]
Anti-TRA-1-60-R Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: TRA-1-60-R]
Supplier:  Biolegend
Certificates
Anti-TRA-1-60-R Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: TRA-1-60-R]
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 330605-BL
  • 330606-BL
  • Antigen Symbol:
  • TRA-1-60-R
  • TRA-1-60-R
  • Antigen Synonyms:
  • Tumor Rejection Antigen 1-60|TRA-1
  • Tumor Rejection Antigen 1-60|TRA-1
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 647
  • Alexa Fluor® 647
  • Clone:
  • TRA-1-60-R
  • TRA-1-60-R
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 5420
  • 5420
  • Isotype:
  • IgM kappa
  • IgM kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Human embryonal carinoma cell line 2102Ep cl.2A6
  • Human embryonal carinoma cell line 2102Ep cl.2A6
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Purification:
  • The antibody was conjugated with Alexa Fluor® 647 under optimal conditions, and is at >85% purity.
  • The antibody was conjugated with Alexa Fluor® 647 under optimal conditions, and is at >85% purity.
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications

Related Information

Frequently Bought Together