Anti-Fc epsilon RI alpha Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: AER-37 (CRA-1)]
Catalog # 334614-BL
Supplier: Biolegend
Specifications
- Antibody Type:Primary
- Antigen Symbol:Fc epsilon RI alpha
- Clonality:Monoclonal
- Clone:AER-37 (CRA-1)
- Conjugation:Alexa Fluor® 647
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Baboon,Cynomolgus monkey,Rhesus monkey,Human
- Size:100 tests
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Gene ID:2205
- Antigen Synonyms:FceRI-a|FceRIa|FceRI alpha|FceRI-alpha|high affinity IgE receptorBasophilEosinophilMast CellPlateletsFcεRIα
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:Ambient RT
- Purification:The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 647 under optimal conditions.
- Cat. No.:334614-BL
- Supplier no.:334614
Specifications
About this item
Alexa Fluor® 647 anti-human FcεRIα [AER-37 (CRA-1)]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: Fc epsilon RI alpha
Clonality: Monoclonal
Clone: AER-37 (CRA-1)
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Baboon;Cynomolgus;Rhesus