Order Entry
United States
ContactUsLinkComponent
Anti-HLA-DR Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: L243]
Anti-HLA-DR Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: L243]
Supplier:  Biolegend
Certificates
Anti-HLA-DR Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: L243]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • 10757-986
  • 307620-BL
  • 307619-BL
  • Antigen Symbol:
  • HLA-DR
  • HLA-DR
  • HLA-DR
  • Antigen Synonyms:
  • Major Histocompatibility Class II|HLADR !HLADRG46-6|MHC class II|G466
  • Major Histocompatibility Class II|HLADR !HLADRG46-6|MHC class II|G466
  • Major Histocompatibility Class II|HLADR !HLADRG46-6|MHC class II|G466
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Clone:
  • L243
  • L243
  • L243
  • Reactivity:
  • Marmoset, Baboon, Tamarin, African green monkey, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Dog, Squirrel monkey
  • Marmoset, Baboon, Tamarin, African green monkey, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Dog, Squirrel monkey
  • Marmoset, Baboon, Tamarin, African green monkey, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Dog, Squirrel monkey
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 3122, 3123
  • 3122, 3123
  • 3122, 3123
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • IgG2a kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Form:
  • µg size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide. Test sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • µg size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide. Test sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • µg size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide. Test sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 488 under optimal conditions.
  • The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 488 under optimal conditions.
  • The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 488 under optimal conditions.
  • Concentration:
  • 0.50 mg/mL
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT
  • Ambient RT

Specifications

Related Information

Frequently Bought Together