Anti-WNT1 Rabbit Polyclonal Antibody
RL600-401-A37
- Antibody Type:Primary
- Antigen Name:Wingless-type MMTV Integration Site Family Member 1
- Antigen Symbol:WNT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Opossum,Macaque,Human,Bovine,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Epitope:Center
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:7471
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage Temperature:–20 °C
- Concentration:1.38 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of human Wnt1 Protein.
- Purification:affinity-purified
- Cat. No.:RL600-401-A37
- Supplier no.:600-401-A37
Type: Primary
Antigen: WNT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Human, Mouse, Rat, macaque, opossum