Order Entry
United States
ContactUsLinkComponent
Anti-IgG Donkey Antibody (Biotin)
Anti-IgG Donkey Antibody (Biotin)
Catalog # 95017-418
Supplier:  Cytiva
Product available on GSA Advantage®
Anti-IgG Donkey Antibody (Biotin)
Catalog # 95017-418
Supplier:  Cytiva
Supplier Number:  RPN1004
Product available on GSA Advantage®

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Conjugation:
    Biotin
  • Host:
    Donkey
  • Isotype:
    IgG
  • Size:
    2 mL
  • Cat. No.:
    95017-418
  • Supplier no.:
    RPN1004

Specifications

About this item

The antibodies are supplied with dilution guidelines for relevant applications.

  • Soluble in cold water and hot water

The antibodies are supplied with dilution guidelines for relevant applications and are stored at 2°C to 8°C.

Type: Secondary
Antigen: Rabbit Ig
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Donkey
Isotype: none
Reactivity: Rabbit