Anti-IgG Sheep Antibody (HRP (Horseradish Peroxidase))
Catalog # 95017-326
Supplier: Cytiva
Product available on GSA Advantage®
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Sheep
- Isotype:IgG
- Reactivity:Human
- Size:1 mL
- Antibody Fragment:Whole Ab
- Storage Buffer:Horseradish peroxidase conjugated antibody is supplied in phosphate buffered saline (sodium phosphate 0.1 M, NaCl 0.1 M) pH 7.5, containing 1% (w/v) bovine serum albumin and an anti-microbial agent
- Storage Temperature:2 - 8 °C
- Immunogen:Human IgG
- Tested Applications:Western blotting
- Cat. No.:95017-326
- Supplier no.:NA933-1ML
Specifications
About this item
Highly species specific HRP-conjugated antibodies optimized for use with Amersham ECL Western Blotting Detection Reagents.
Optimized for use with our range of Amersham ECL Western blotting detection reagents.
The antibodies are supplied with dilution guidelines for relevant applications and should be stored at 2°C to 8°C.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: none
Reactivity: Human